Learn More
Globe Scientific Quick-Prep™ Urinalysis Kit
Tube and pipette system that isolates 1mL of urine sediment for microscopic examination
Supplier:  Globe Scientific 3810
 View more versions of this product
Catalog No. 21200214
Only null left
Includes:
  • Quick-Prep Urinalysis System (Pipette and Tube) - Transfer pipette and 12mL urine centrifuge tube
  • Sternheimer Malbin Urine Sediment Stain
Description
Description
  • Designed for use with our Quick-Read (10-place) urinalysis slide
Specifications
Specifications
Stemheimer Malbin Urine Sediment Stain
Quick-Read (10-place) urinalysis slide
50 mL
Product Suggestions
Product Suggestions