missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Quick-Prep™ Urinalysis Kit

Numéro de catalogue. p-4660213
Change view
Click to view available options
Quantity:
1000 Tests
50 mL
Type:
Stemheimer Malbin Urine Sediment Stain
Urinalysis Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Type
21200215 1000 Tests Urinalysis Kit
21200214 50 mL Stemheimer Malbin Urine Sediment Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 21200215 Fournisseur Globe Scientific Code fournisseur 3825
Il en reste null

Comprend

  • Quick-Prep Urinalysis System (Pipette and Tube) - Transfer pipette and 12mL urine centrifuge tube
  • Sternheimer Malbin Urine Sediment Stain

Tube and pipette system that isolates 1mL of urine sediment for microscopic examination

  • Designed for use with our Quick-Read (10-place) urinalysis slide

Spécifications

Type Urinalysis Kit
Includes Transfer pipette and 12mL Urine centrifuge tube
Quantity 1000 Tests