Learn More
Globe Scientific Flanged Plug Tube Caps
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
Supplier: Globe Scientific 118127Y
Description
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
Order Info
1000/pk
Chemical Identifiers
| Flange Plug Cap | |
| Polyethylene |
Specifications
| Flange Plug Cap | |
| 0.47 in. | |
| 12 mm Tubes |
| Polyethylene | |
| 12 mm |