Learn More
Globe Scientific Accessories for Spectrophotometer Cuvettes
For use with Spectophotometer cuvettes
Supplier:  Globe Scientific 111168
 View more versions of this product
Catalog No. 21200261
Only null left
Description
Description
  • Polyethylene made 12-place cuvette holder of 170 x 50 x 15mm dimensions
  • LDPE snap caps for square cuvettes for added protection against spillage and contamination
  • Polystyrene disposable 90mm stirring stick for access to the bottom of cuvettes
Specifications
Chemical Identifiers
Spectrophotometer cuvettes
Specifications
Cuvette Holder
Spectrophotometer cuvettes
12 Spectrophotometer cuvettes
Polyethylene
Product Suggestions
Product Suggestions