missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Globe Scientific Accessories for Spectrophotometer Cuvettes
Description
- Polyethylene made 12-place cuvette holder of 170 x 50 x 15mm dimensions
- LDPE snap caps for square cuvettes for added protection against spillage and contamination
- Polystyrene disposable 90mm stirring stick for access to the bottom of cuvettes
Specifications
Specifications
| Type | Plug Cap |
| For Use With (Equipment) | Spectrophotometer cuvettes |
| Material | LDPE |
| Pathlength | 10 mm |