Learn More
Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes, Yellow
Conform well to irregular, curved, rough and highly textured surfaces. Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes are perfect for harsh, industrial environments.
Supplier:  Brady™ 113115
 View more versions of this product
Catalog No. 19107942
Description
Description
  • Adhere to difficult low surface energy items
  • 5 year outdoor durability
  • Sticks to high textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes
Specifications
Chemical Identifiers
BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863
Acrylic, Permanent, Cold-temperature
Yellow
Continuous
Thermal Transfer
Arc Flash Labels, Arrow Tape, Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking
1
1 Roll/Box
-40°C to +82°C
0.102 mm
30.48 m
Suggested Ribbon Series: R10000
None
Gloss
Vinyl Film (B-595)
Non-sterile
GlobalMark™ Label Maker
-40°C
-40°F to +180°F
0.004 in.
100 ft.
Specifications
BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863
Acrylic, Permanent, Cold-temperature
Yellow
Continuous
Single-side Printable
Tape
Arc Flash Labels, Arrow Tape, Pipe and Valve Marking, Safety and Compliance ID, Visual Workplace Labeling, Warehouse Marking
1
1 Roll/Box
-40°C to +82°C
0.102 mm
100 ft.
3 in.
3 in.
Suggested Ribbon Series: R10000
None
Gloss
Vinyl Film (B-595)
Thermal Transfer
Non-sterile
GlobalMark™ Label Maker
-40°C
-40°F to +180°F
0.004 in.
30.48 m x 76.2 mm
30.48 m
76.20 mm
Product Suggestions
Product Suggestions