Learn More
Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes, Clear
Conform well to irregular, curved, rough and highly textured surfaces. Brady™ GlobalMark™ Indoor/Outdoor Grade Vinyl (B-595) Tapes are perfect for harsh, industrial environments.
Supplier:  Brady™ 113107
 View more versions of this product
Catalog No. 19107939
Only null left
Description
Description
  • Adhere to difficult low surface energy items
  • 5 year outdoor durability
  • Sticks to high textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes
Specifications
Chemical Identifiers
Suggested Ribbon Series: R10000
Clear
Tape
GlobalMark™ Label Maker
100 ft.
Acrylic, Permanent, Cold-temperature
Vinyl Film (B-595)
Arc Flash Labels; Arrow Tape; Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking
0.004 in.
Specifications
Suggested Ribbon Series: R10000
Clear
Tape
GlobalMark™ Label Maker
100 ft.
Acrylic, Permanent, Cold-temperature
Vinyl Film (B-595)
Arc Flash Labels; Arrow Tape; Pipe and Valve Marking; Safety and Compliance ID; Visual Workplace Labeling; Warehouse Marking
0.004 in.
1.125 in.
Product Suggestions
Product Suggestions