missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLO1 Recombinant Rabbit Monoclonal Antibody (23GB5555)
Rabbit Recombinant Monoclonal Antibody
$656.94 - $1779.35
Specifications
| Antigen | GLO1 |
|---|---|
| Clone | 23GB5555 |
| Concentration | 0.2 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
GLO1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere.Specifications
| GLO1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q04760, Q6P7Q4, Q9CPU0 | |
| 109801, 2739, 294320 | |
| A synthesized peptide derived from human GLO1 (10-50AA). | |
| Antibody |
| 23GB5555 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GLO1 | |
| 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; aldoketomutase; AW550643; cb554; epididymis secretory protein Li 74; GLO1; Glo-1; Glo-1r; Glo1-r; Glo-1s; Glo1-s; GLOD1; glx I; GLY1; GLYI; glyoxalase 1; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; glyoxalase I; glyoxylase 1; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; lactoylglutathione lyase GLO1; methylglyoxalase; Qglo; S-D-lactoylglutathione methylglyoxal lyase; wu:fb82g09; YM9571.15C; YML004C; zgc:66035 | |
| GLO1 | |
| Primary | |
| Affinity chromatography |