missing translation for 'onlineSavingsMsg'
Learn More

Gliquidone 98.0+%, TCI America™

Catalog No. p-7130242
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03321G 1 g
G03325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03321G Supplier TCI America Supplier No. G03321G
Only null left

Chemical Identifiers

CAS 33342-05-1
Molecular Formula C27H33N3O6S
Molecular Weight (g/mol) 527.636
MDL Number MFCD00631870
InChI Key LLJFMFZYVVLQKT-UHFFFAOYSA-N
PubChem CID 91610
IUPAC Name 1-cyclohexyl-3-[4-[2-(7-methoxy-4,4-dimethyl-1,3-dioxoisoquinolin-2-yl)ethyl]phenyl]sulfonylurea
SMILES CC1(C2=C(C=C(C=C2)OC)C(=O)N(C1=O)CCC3=CC=C(C=C3)S(=O)(=O)NC(=O)NC4CCCCC4)C

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 1 g
Formula Weight 527.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Gliquidone
RTECSNumber DB1584250
TSCA No