missing translation for 'onlineSavingsMsg'
Learn More

Glipizide

Catalog No. AAJ6339803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6339803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6339803 Supplier Thermo Scientific Chemicals Supplier No. J6339803

An ATP-dependent potassium channel blocker | CAS: 29094-61-9 | C21H27N5O4S | 445.538 g/mol

An ATP-dependent potassium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 29094-61-9
Color White to Yellow
Molecular Formula C21H27N5O4S
Quantity 1 g
MDL Number MFCD00072159
Merck Index 14,4442
Synonym glipizide, glucotrol, glydiazinamide, melizide, glibenese, glucozide, glupizide, sucrazide, dipazide, glupitel
Solubility Information Soluble in methanol (1.9mg/ml) and DMSO (48mg/ml)
InChI Key ZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILES CC1=NC=C(N=C1)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3
IUPAC Name N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-5-methylpyrazine-2-carboxamide
Molecular Weight (g/mol) 445.538
PubChem CID 3478
ChEBI CHEBI:5384
Formula Weight 445.54
Physical Form Powder
Chemical Name or Material Glipizide
Show More Show Less
EINECSNumber 249-427-6
RTECSNumber YS7640000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only