missing translation for 'onlineSavingsMsg'
Learn More

Glipizide Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013945
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013945 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013945 Supplier Merck Emd Millipore Supplier No. PHR181350MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glipizide is an oral antidiabetic drug that belongs to the class of second-generation sulfonylureas. It is used in the treatment of type 2 diabetes mellitus. It exerts its action by lowering the blood glucose levels via stimulation of insulin release from the β-cells of the pancreas. Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 33288-71-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C14H16N4O3 S
Synonym N-{2-[4-(Aminosulfonyl)phenyl]ethyl}-5-methyl-2-pyrazinecarboxamide; N-{2-[(4-aminosulfonyl)phenyl]ethyl}-5-methyl-pyrazinecarboxamide; 4-[2-(5-Methylpyrazine-2-carboxamido)ethyl]benzenesulfonamide
Type Certified Reference Material
Quantity 50 mg
Formula Weight 320.37
Grade Certified Reference Material