missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012715
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012715 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012715 Supplier Merck Emd Millipore Supplier No. PHR172740MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Pharmaceutical (small molecule)

Specifications

CAS 119018-30-3
Linear Formula C18H23N3O6 S
Synonym N-[[4-[2-[[(3-Ethyl-2,5-dihydro-4-methyl-2-oxo-1 H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]carbamic acid methyl ester; Glimepiride Urethane; Methyl ((4-(2-(3-ethyl-4-methyl-2-oxo-2,5-dihydro-1 H-pyrrole-1-carboxamido)ethyl)phenyl)sulfonyl)carbamate
Type Certified Reference Material
Quantity 40 mg
Formula Weight 409.46
Grade Certified Reference Material