missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride cis-Isomer (Glimepiride Related Compound A) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015170
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015170 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015170 Supplier Merck Emd Millipore Supplier No. PHR172540MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glimepiride cis-Isomer (Glimepiride Related Compound A) is an impurity of glimepiride. Glimepiride is a third generation, medium-to-long acting sulphonylurea compound used as an oral antidiabetic drug. Its mode of action involves the lowering of blood glucose by stimulating the release of insulin from functioning pancreatic beta cells.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 684286-46-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C24H34N4O5 S
Synonym 3-Ethyl-2,5-dihydro-4-methyl-N-[2-[4-[[[[(cis-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1 H-pyrrole-1-carboxamide; 1-[[p-[2-(3-Ethyl-4-methyl-2-oxo-3-pyrroline-1-carboxamido)ethyl]phenyl]sulfonyl]-3-(cis-4-methylcyclohexyl)urea; Glimepiride cis-isomer
Type Certified Reference Material
Quantity 40 mg
Formula Weight 490.62
Grade Certified Reference Material