missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 461070050

 View more versions of this product

Catalog No. AC461070050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Gliclazide
0.5% max. (1 g, 105°C)
C15H21N3O3S
5g
Solubility in water: insoluble. Other solubilities: soluble in dmf, slightly soluble in methanol, ethanol
CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1
323.41
CHEBI:31654
≥98.5% (HPLC)
21187-98-4
Authentic
MFCD00409893
gliclazide, glimicron, diamicron, nordialex, diaikron, gliclazida, gliklazid, glyclazide, gliclazidum inn-latin, gliclazida inn-spanish
BOVGTQGAOIONJV-UHFFFAOYNA-N
1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(4-methylphenyl)sulfonylurea
3475
323.41

Chemical Identifiers

21187-98-4
323.41
BOVGTQGAOIONJV-UHFFFAOYNA-N
3475
1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(4-methylphenyl)sulfonylurea
C15H21N3O3S
MFCD00409893
gliclazide, glimicron, diamicron, nordialex, diaikron, gliclazida, gliklazid, glyclazide, gliclazidum inn-latin, gliclazida inn-spanish
CHEBI:31654
CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Suspected of damaging fertility or the unborn child.

GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 244-260-5

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only