missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide 98.5+%, TCI America™

Catalog No. g03815g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
G03815G 5 g
1 options

Catalog No. G03815G

Supplier: TCI America G03815G

Only null left

Chemical Identifiers

CAS 21187-98-4
Molecular Formula C15H21N3O3S
Molecular Weight (g/mol) 323.41
MDL Number MFCD00409893
InChI Key BOVGTQGAOIONJV-UHFFFAOYNA-N
Synonym gliclazide, glimicron, diamicron, nordialex, diaikron, gliclazida, gliklazid, glyclazide, gliclazidum inn-latin, gliclazida inn-spanish
PubChem CID 3475
ChEBI CHEBI:31654
IUPAC Name 1-(4-methylbenzenesulfonyl)-3-{octahydrocyclopenta[c]pyrrol-2-yl}urea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1

Specifications

Melting Point 167°C
Color White
Quantity 5 g
Formula Weight 323.41
Percent Purity ≥98.5% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Gliclazide
RTECSNumber YT4500000
TSCA No