missing translation for 'onlineSavingsMsg'
Learn More

Glibenclamide 98.5+%, TCI America™

Catalog No. p-7130252
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03825G 5 g
G038225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03825G Supplier TCI America Supplier No. G03825G

Chemical Identifiers

CAS 10238-21-8
Molecular Formula C23H28ClN3O5S
Molecular Weight (g/mol) 494.003
MDL Number MFCD00056625
InChI Key ZNNLBTZKUZBEKO-UHFFFAOYSA-N
Synonym glyburide, glibenclamide, glybenclamide, micronase, diabeta, glynase, daonil, euglucon, maninil, semi-daonil
PubChem CID 3488
ChEBI CHEBI:5441
IUPAC Name 5-chloro-N-[2-[4-(cyclohexylcarbamoylsulfamoyl)phenyl]ethyl]-2-methoxybenzamide
SMILES COC1=C(C=C(C=C1)Cl)C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC3CCCCC3

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 5 g
Formula Weight 494.00
Percent Purity ≥98.5% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glibenclamide
EINECSNumber (9)-0412
RTECSNumber YS4725200
TSCA No
Recommended Storage Refrigerator