missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GLG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526838
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526838 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526838 Supplier Invitrogen™ Supplier No. PA526838
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

This antibody is predicted to react with chicken, hamster and rat based on sequence homology.

GLG1 is a protein coding gene. Binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils).

Specifications

Antigen GLG1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GLG1
Gene Accession No. Q61543, Q92896
Gene Alias AI593353; AW537898; CFR; CFR1; CFR-1; Cysteine-rich fibroblast growth factor receptor; E-selectin ligand 1; ESL1; ESL-1; Glg1; Golgi apparatus protein 1; golgi glycoprotein 1; golgi sialoglycoprotein MG-160; MG160; MG-160; selectin, endothelial cell, ligand; Selel
Gene Symbols GLG1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1152-1179 amino acids from the C-terminal region of human GLG1.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20340, 2734
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less