missing translation for 'onlineSavingsMsg'
Learn More

GlcNAc beta(1-3)[Neu5Ac alpha(2-6)]GalNAc-alpha-pNP 97.0+%, TCI America™

Catalog No. G03782MG
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03782MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03782MG Supplier TCI America Supplier No. G03782MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1858223-99-0
Molecular Formula C33H48N4O21
Molecular Weight (g/mol) 836.754
InChI Key LQCFVBAMHWMDAT-PQEAEXJMSA-N
PubChem CID 121235354
IUPAC Name (2R,4S,5R,6R)-5-acetamido-2-[[(2R,3R,4R,5R,6R)-5-acetamido-4-[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-hydroxy-6-(4-nitrophenoxy)oxan-2-yl]methoxy]-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OCC2C(C(C(C(O2)OC3=CC=C(C=C3)[N+](=O)[O-])NC(=O)C)OC4C(C(C(C(O4)CO)O)O)NC(=O)C)O)O

Specifications

Color White-Yellow
Quantity 2 mg
Formula Weight 836.75
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNAc beta(1-3)[Neu5Ac alpha(2-6)]GalNAc-alpha-pNP
TSCA No
Recommended Storage Freezer