missing translation for 'onlineSavingsMsg'
Learn More

GlcNAc beta(1-3)[GlcNAc beta(1-6)]GalNAc-alpha-Thr 94.0+%, TCI America™

Catalog No. G03422MG
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03422MG 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03422MG Supplier TCI America Supplier No. G03422MG
Only null left

Chemical Identifiers

CAS 1304646-03-4
Molecular Formula C28H48N4O18
Molecular Weight (g/mol) 728.702
InChI Key HWIHOSGHOVCLLL-NQHHJXOOSA-N
PubChem CID 53384352
IUPAC Name (2S,3R)-3-[(2S,3S,4R,5R,6S)-3-acetamido-4-[(2R,3S,4R,5S,6S)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[[(2R,3S,4R,5S,6S)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]-5-hydroxyoxan-2-yl]oxy-2-aminobutanoic acid
SMILES CC(C(C(=O)O)N)OC1C(C(C(C(O1)COC2C(C(C(C(O2)CO)O)O)NC(=O)C)O)OC3C(C(C(C(O3)CO)O)O)NC(=O)C)NC(=O)C

Specifications

Color White
Quantity 2 mg
Formula Weight 728.70
Percent Purity ≥94.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material GlcNAc beta(1-3)[GlcNAc beta(1-6)]GalNAc-alpha-Thr
TSCA No
Recommended Storage Freezer