CAS RN 1304646-03-4
CAS RN 1304646-03-4
IUPAC Name:
2-amino-3-[(3-acetamido-4-{[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-({[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)-5-hydroxyoxan-2-yl)oxy]butanoic acid
Synonyms:
2-amino-3-[(3-acetamido-4-{[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-6-({[3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}methyl)-5-hydroxyoxan-2-yl)oxy]butanoic acid
Molecular Weight (g/mol):
728.7
Molecular Formula:
C28H48N4O18
InChi Key:
HWIHOSGHOVCLLL-UHFFFAOYNA-N
SMILES:
CC(OC1OC(COC2OC(CO)C(O)C(O)C2NC(C)=O)C(O)C(OC2OC(CO)C(O)C(O)C2NC(C)=O)C1NC(C)=O)C(N)C(O)=O
1
–
1
of
1
results
1
GlcNAc beta(1-3)[GlcNAc beta(1-6)]GalNAc-alpha-Thr 94.0+%, TCI America™
CAS: 1304646-03-4 Molecular Formula: C28H48N4O18 Molecular Weight (g/mol): 728.702 InChI Key: HWIHOSGHOVCLLL-NQHHJXOOSA-N PubChem CID: 53384352 IUPAC Name: (2S,3R)-3-[(2S,3S,4R,5R,6S)-3-acetamido-4-[(2R,3S,4R,5S,6S)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[[(2R,3S,4R,5S,6S)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]-5-hydroxyoxan-2-yl]oxy-2-aminobutanoic acid SMILES: CC(C(C(=O)O)N)OC1C(C(C(C(O1)COC2C(C(C(C(O2)CO)O)O)NC(=O)C)O)OC3C(C(C(C(O3)CO)O)O)NC(=O)C)NC(=O)C