Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10
Precision manufactured, 10-chambered slide available without a hemocytometer-type grid
$260.00 - $492.64
Specifications
| Type | Slide 10 |
|---|---|
| For Use With (Application) | Microscopy |
| Gridded | No |
| Material | Plastic with the optical qualities of glass |
| No. of Wells | 10 |
Description
Glasstic Slides sets the standard for accurate, reproducible, cost-effective and time-efficient standardized microscopic analysis.
- Made from optically clear plastic
- Glasstic Slide features 10 individual fully segregated specimen wells
- Capillary action reduces the likelihood of pipetting errors
- Suspends sediments homogeneously throughout the specimen
- Eliminates other variables, providing precise, reproducible fluid distribution throughout each chamber
- Reproducible fluid distribution throughout each chamber
- Chamber walls prevent specimen leakage and cross-contamination
- Correlate well with results obtained by using a hemocytometer: red blood cells: r: 0.99; white blood cells: r: 0.99; platelets: r: 0.93
Specifications
| Slide 10 | |
| No | |
| 10 |
| Microscopy | |
| Plastic with the optical qualities of glass |