missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva Glass Vacuum Filtration Device, Upper Portion

Produced from borosilicate glass and suitable for aqueous and organic solvent filtration. The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane.

Supplier:  Cytiva 10443002

 View more versions of this product

Catalog No. 09927395



Description

Description

  • Chemically resistant to most aqueous and organic solutions
  • Resistant to acids and caustic solutions
  • Autoclavable and can be sterilized in dry heat to 180°C
  • Can be used up to 200°C
Note: Erlenmeyer Flask (NS45) is sold separately.

Specifications

Specifications

Hose coupling connection
70 cm2
Borosilicate glass
Upper portion of glass vacuum filtration device
225 mm
Borosilicate Glass with Stainless Steel Clamp
Filtration and biological analysis, the 304 Stainless Steel support screen is suitable for use with proteinaceous solutions
1 Ea.
500 mL (Funnel)
90 mm
SDS
Documents

Documents

Promotions

Promotions