missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glass Spheres , MilliporeSigma™ Supelco™
Silica is a naturally occurring material in minerals, flint and in some plants in crystalline phase. Silica used in industries is in synthetic form. Surface area, pore volume, pore size and particle size are independently controllable to some extent. The crystalline silica may be classified based on atmospheric pressure as:a. Quartz = 1143Kb. Tridymite 1143 - 1743Kc. Cristobalite = 1743, over 1973K it forms amorphous vitreous silica glass.
$75.51 - $208.01
Specifications
| CAS | 65997-17-3 |
|---|---|
| Density | 1.1 g/mL (at 25°C) |
| Packaging | Poly Bottle |
| Linear Formula | O2Si |
| MDL Number | MFCD00147032 |
Specifications
| 65997-17-3 | |
| Poly Bottle | |
| MFCD00147032 | |
| High-Purity Grade |
| 1.1 g/mL (at 25°C) | |
| O2Si | |
| 60.08 | |
| Powder |