missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Reagent Bottles with Polypropylene Stopper

Catalog No. S39281
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
500 mL
2 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S39281 30 mL
S39282 60 mL
S39283 125 mL
S39285 500 mL
S39287 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S39281 Supplier Eisco™ Supplier No. CH0160A

Includes

Clear glass bottle, polypropylene stopper

Clear glass reagent bottles with polypropylene stoppers are ideal for storing liquid solutions in the laboratory. The polypropylene stoppers are acid proof. Both the glass bottles and stoppers are autoclavable.

  • Made of high quality borosilicate 3.3 glass
  • Narrow mouth with acid proof polypropylene stopper
  • Bottles and stoppers are autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (English) 1.3 in.
For Use With (Application) Solution Preparation, Solution Storage
Includes Clear glass bottle, polypropylene stopper
Material Borosilicate Glass
Length (Metric) 33mm
Quantity 1
Sterility Non-sterile
Weight (Metric) 45.35 g
Width (Metric) 33mm
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Cylindrical
Height (Metric) 81.3 mm
Product Type Reagent Bottle
For Use With (Equipment) Liquids
Height (English) 3.2 in.
Chemical Label Unlabeled
Closure Material Polypropylene
Closure Size 14/23 mm
Diameter (Metric) 33 mm
Closure Type Stopper
Autoclavable Autoclavable
Show More Show Less