missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Reagent Bottles with Hollow Interchangeable Hexagonal Glass Stoppers

Clear glass reagent bottles with interchangeable hollow glass stoppers are well suited for storing liquid solutions in the laboratory. Glass bottles and glass stoppers are autoclavable.

Supplier:  Eisco™ CH0162F

 View more versions of this product

Catalog No. S39293


Only null left

Includes: Clear glass bottle, interchangeable hollow hexagonal glass stopper

Description

Description

  • Made of high quality borosilicate 3.3 glass
  • Narrow mouth with interchangeable hollow hexagonal glass stopper
  • Bottle and stopper are autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

1 L
Clear
4 in.
Interchangeable hexagonal glass hollow stopper
101.6mm
Non-sterile
101.6mm
Yes
No
Cylindrical
Reagent Bottle
9 in.
Glass
101.6 mm
Autoclavable
33.8 oz.
Hollow stopper
Solution Preparation, Solution Storage
Borosilicate Glass
1
408.23 g
Clear
Reusable
Narrow
228.6 mm
Liquids
Unlabeled
29/32 mm
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions