missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Reagent Bottles with Hollow Interchangeable Hexagonal Glass Stoppers

Clear glass reagent bottles with interchangeable hollow glass stoppers are well suited for storing liquid solutions in the laboratory. Glass bottles and glass stoppers are autoclavable.

Supplier:  Eisco™ CH0162E

 View more versions of this product

Catalog No. S39292


Only null left

Includes: Clear glass bottle, interchangeable hollow hexagonal glass stopper

Description

Description

  • Made of high quality borosilicate 3.3 glass
  • Narrow mouth with interchangeable hollow hexagonal glass stopper
  • Bottle and stopper are autoclavable

Warranty

1 year limited manufacturer warranty

Specifications

Specifications

500 mL
Clear
2.5 in.
Interchangeable hexagonal glass hollow stopper
63.5mm
Non-sterile
63.5mm
Yes
No
Cylindrical
Reagent Bottle
5.74 in.
Glass
63.5 mm
Autoclavable
16.9 oz.
Hollow stopper
Solution Preparation, Solution Storage
Borosilicate Glass
1
317.51 g
Clear
Reusable
Narrow
146 mm
Liquids
Unlabeled
24/29 mm
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions