Learn More
Cytiva Glass Plate
Description
- Glass Plates
- For use with SE 600 Vertical Unit
- Dimensions: 18 x 24 cm
- Pack Size: 2
Specifications
Specifications
| For Use With (Equipment) | SE 600 Vertical Unit |
| Type | Glass Plate |
| Material | Glass |
| Quantity | 2/Pk. |
| Dimensions (L x W) | 18 x 24 cm |