Learn More
Eisco™ Glass Plano Convex Lenses
These glass plano-convex lenses feature one flat side and one convex side, ideal for physical science demonstration and experiments
Supplier:  Eisco™ PH0532ECV
 View more versions of this product
Catalog No. S27387
Only null left
Description
Description
  • High quality, clear glass
  • Optically worked
  • Highly polished
  • Spherical
Specifications
Chemical Identifiers
Glass
2 in.
Plano
50 mm
Physical Science Demonstration and Experiments
Specifications
Glass
2 in.
500 mm
Convex Circle
50 mm
Physical Science Demonstration and Experiments
Plano
Product Suggestions
Product Suggestions