Learn More
MilliporeSigma™ Glass Funnel for Vacuum Filtration
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Supplier:  MilliporeSigma™ XX1014707
 View more versions of this product
Catalog No. XX1014707
Only null left
Description
Description
Designed to replace parts contained in glass vacuum filtration kits or provide for an alternative configuration
Millipore Glass Vacuum Filtration Accessory ProductThese accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration. This item is an accessory part for the Millipore 47 mm Classic Glass Filter Holder Kits, Millipore 47 mm All-Glass Filter Holder Kits, and Millipore MilliSolve Kit.
Specifications
Chemical Identifiers
47 mm
Clear
Specifications
47 mm
Borosilicate Glass
Ground Glass Seal
1 L
Clear
Product Suggestions
Product Suggestions