Learn More
Eisco™ Glass Fractional Distillation Column
Description
- Fractional distillation column made from high quality borosilicate glass
- Designed with a rod and disc pattern.
- Includes a removable center rod with 20 discs fit inside a smooth bore tube with side arm
- Overall length is 480 mm
Warranty
1 year limited manufacturer warranty
Specifications
Specifications
| Height (Metric) | 480 mm |
| For Use With (Application) | Distilation |
| Type | Round and disk pattern |
| Material | Borosilicate Glass |
| Quantity | 1 |