Learn More
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals
Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.
Supplier:  DWK Life Sciences 7472051320
 View more versions of this product
Catalog No. K7472051320
Only null left
Description
Description
  • Connectors have PTFE seals and FKM O-Rings
  • Blue glass-filled nylon
  • For use to 150°C
Specifications
Chemical Identifiers
Nylon
Connector
To connect standard rotary evaporator traps to vials
PTFE seals
Specifications
Connector
13-425 to 20-400
PTFE seals
To connect standard rotary evaporator traps to vials
Nylon
Product Suggestions
Product Suggestions