Learn More
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals
Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.
Supplier: DWK Life Sciences 7472051315
Description
- Connectors have PTFE seals and FKM O-Rings
- Blue glass-filled nylon
- For use to 150°C
Chemical Identifiers
| Nylon | |
| Connector | |
| To connect standard rotary evaporator traps to vials | |
| PTFE seals |
Specifications
| To connect standard rotary evaporator traps to vials | |
| Connector | |
| Nylon |
| 13-425 to 15-425 | |
| PTFE seals |