missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Clear Glass Dropping Bottles

Can be used to store and dispense liquids.

$9.54 - $17.43

Specifications

Color Clear
For Use With (Application) For transferring and mixing essential oils, indicators, stains or other aqueous solutions
Includes Dropper
Material Soda Lime Glass
Quantity 1 Ea.
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S28016
View Documents
Eisco™
CH0187A
30 mL
Each for $9.54
Only null left
 
S28017
View Documents
Eisco™
CH0187B
60 mL
Each for $10.64
Only null left
 
S28018
View Documents
Eisco™
CH0187C
100 mL
Each for $17.43
Only null left
 
Description

Description

  • Made of Soda Glass
  • With screw cap and fitted with dropper
Specifications

Specifications

Clear
Dropper
1 Ea.
Non-sterile
Yes
No
Dropping Bottle
Glass/Rubber
Autoclavable
For transferring and mixing essential oils, indicators, stains or other aqueous solutions
Soda Lime Glass
Black
Red
Reusable
Cylindrical
Blank
Screw Cap
Videos
SDS
Documents

Documents