missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Glass Desiccator with Porcelain Plate

Typically used for the storage of hygroscopic, moisture sensitive chemicals and reagents.

Supplier:  Eisco™ CH0362B

 View more versions of this product

Catalog No. S27342


Only null left

Description

Description

  • Includes glass cover with knob and unglazed porcelain plate
  • Constructed of high quality, heavy duty 3.3 borosilicate glass
  • Eisco Labs Borosilicate glass has the following heat tolerances:
    • Strain Point: -515°C
    • Annealing Point: 565°C
    • Softening Point: 820°C
Specifications

Specifications

Borosilicate Glass (3.3)
Porcelain Plate
11 x 10.5 in.
26.7 cm
11 in.
10.5 in.
13 mm
Desiccator with Lid
1 Ea.
10.5 in.
28 cm
7.5 in.
19 cm
15 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions