missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Desiccator with Porcelain Plate
Typically used for the storage of hygroscopic, moisture sensitive chemicals and reagents.
$633.07 - $720.43
Specifications
| Material | Borosilicate Glass (3.3) |
|---|---|
| Product Type | Desiccator with Lid |
| Includes | Porcelain Plate |
| Quantity | 1 Ea. |
Description
- Includes glass cover with knob and unglazed porcelain plate
- Constructed of high quality, heavy duty 3.3 borosilicate glass
- Eisco Labs Borosilicate glass has the following heat tolerances:
- Strain Point: -515°C
- Annealing Point: 565°C
- Softening Point: 820°C
Specifications
| Borosilicate Glass (3.3) | |
| Desiccator with Lid | |
| Porcelain Plate | |
| 1 Ea. |