missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp™ Glass Crucible P2
Description
Holds samples for general laboratory purposes. Velp™ Glass Crucible P2 is an excellent accessory for Velp™ FIWE Series Fiber Analyzers.
- Durable glass
- Standardized
Specifications
Specifications
| For Use With (Equipment) | Fiber Analyzers (FIWE Series) |
| Product Type | Crucible |
| Material | Glass |
| Sterility | Sterile |
| Color | Clear |
| Quantity | 6/Pk. |
| Frequency | 50/60 Hz |
| Voltage | 115/230 VAC |