missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Glass Convex Mirror
Round, optically worked Glass Convex Mirror with a silvered back and protective coating. Designed for physical science demonstrations and experiments.
$8.54 - $93.54
Specifications
| Material | Glass |
|---|---|
| For Use With (Application) | Physical Science Demonstrations and Experiments |
| Product Type | Convex Mirror |
| Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) Focal | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) Focal | Price | Quantity & Availability | ||||
|
S27367
|
Eisco™
PH0521A |
38 mm | 50 mm |
Each for $11.79
|
|
||||
|
S27368
|
Eisco™
PH0521B |
38 mm | 100 mm |
Each for $8.54
|
|
||||
|
S27369
|
Eisco™
PH0521C |
38 mm | 150 mm |
Each for $8.54
|
|
||||
|
S27370
|
Eisco™
PH0521D |
38 mm | 200 mm |
Each for $8.54
|
|
||||
|
S27697
|
Eisco™
PH0526A |
50 mm | 50 mm |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
S27371
|
Eisco™
PH0526B |
50 mm | 75 mm |
Each for $8.54
|
|
||||
|
S27698
|
Eisco™
PH0526C |
50 mm | 100 mm |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
S27699
|
Eisco™
PH0526D |
50 mm | 150 mm |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
S27700
|
Eisco™
PH0526E |
50 mm | 200 mm |
Each for $13.00
|
|
||||
|
S27372
|
Eisco™
PH0526F |
50 mm | 300 mm |
Each for $8.54
|
|
||||
|
S27373
|
Eisco™
PH0526H |
75 mm | 75 mm |
Each for $18.68
|
|
||||
|
S27374
|
Eisco™
PH0526I |
75 mm | 100 mm |
Each for $17.89
|
|
||||
|
S27375
|
Eisco™
PH0526J |
75 mm | 150 mm |
Each for $17.89
|
|
||||
|
S27376
|
Eisco™
PH0526K |
75 mm | 200 mm |
Each for $17.89
|
|
||||
|
S27377
|
Eisco™
PH0526L |
75 mm | 300 mm |
Each for $17.89
|
|
||||
|
S83610
|
Eisco™
PH0526M |
100 mm | 50 mm |
Each for $38.18
|
|
||||
|
S27378
|
Eisco™
PH0526N |
100 mm | 100 mm |
Each for $38.18
|
|
||||
|
S27379
|
Eisco™
PH0526O |
150 mm | 150 mm |
Each for $93.54
|
|
||||
|
S83611
|
Eisco™
PH0526P |
150 mm | 300 mm |
Each for $93.54
|
|
||||
Description
- Silvered back with protective coating
- Designed for physics and educational uses
- Mirrors may have minor imperfections that do not affect their physical function
Specifications
| Glass | |
| Physical Science Demonstrations and Experiments | |
| Convex Mirror |