Learn More
Eisco™ Glass Concave Mirror
Round, optically worked Glass Concave Mirror with a silvered back and protective coating. Designed for physical science demonstrations and experiments.
Supplier:  Eisco™ PH0520B
 View more versions of this product
Catalog No. S27363
Only null left
Description
Description
  • For educational use only
Specifications
Chemical Identifiers
Glass
Physical Science Demonstrations and Experiments
Mirror
Specifications
Glass
2 in.
3 in.
Mirror
50 mm
Physical Science Demonstrations and Experiments
75 mm
Concave Circle
Product Suggestions
Product Suggestions