missing translation for 'onlineSavingsMsg'
Learn More

Glacial Acetic Acid, USP, 99.5-100.5%, Spectrum™ Chemical

Catalog No. 18602915
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
Poly-Coated Glass Bottle
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602915 4 x 4 L Poly-Coated Glass Bottle
18602907 100 mL Amber Glass Bottle
18602908 500 mL Amber Glass Bottle
18602911 2.5 L Glass Bottle
18602910 2.5 L Poly-Coated Glass Bottle
18602909 6 x 500 mL Amber Glass Bottle
18602912 4 L Poly-Coated Glass Bottle
18602913 4 x 2.5 L Glass Bottle
18602914 4 x 2.5 L Poly-Coated Glass Bottle
18602916 20 L Poly Drum
18602917 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18602915 Supplier Spectrum Chemical Mfg Cor Supplier No. AC1104LTPVCS4
Only null left

AC110, 64-19-7, C2H4O2

Spectrum™ Chemical Glacial Acetic Acid, USP does have some antibacterial and antifungal properties but is used primarily as a pH adjuster. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Max % 100%
UN Number UN2789
Percent Purity 99.5 to 100.5%
Grade USP
Quantity 4 x 4 L
Packaging Poly-Coated Glass Bottle