missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GK2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002712B01P
Description
Sequence: MAAPKTAAVGPLVGAVVQGTNSTRFLVFNSKTAELLSHHKVELTQEFPKEGWVEQDPKEILQSVYECIARTCEKLDELNIDISNIKAVGVSNQRETTVIWDKLTGEPLYNAVVWLDLRTQTTVEDLSKKIPGNSNFVKSKTGLPLSTYFSAVKLRWMLDNVRNVQKAVEEGRALFGTIDSWLIWSLTGGVNGGVHCTDVTNASRTMLFNIHSLEWDKELCDFFEIPMDLLPNVFSSSEIYGLIKTGALEGVPISGCLGDQCAALVGQMCFQEGQAKNTYGTGCFLLCNTGRKCVFSEHGLLTTVAYKLGREKPAYYALEGSVAIAGAVIRWLRDNLGIIETSGDIERLAKEVGTSYGCYFVPAFSGLYAPYWEPSARGILCGLTQFTNKCHIAFAALEAVCFQTREILEAMNRDCGIPLRHLQVDGGMTNNKVLMQLQADILHIPVIKPFMPETTALGAAMAAGAAEGVSVWSLEPQALSVLRMERFEPQIQATESEIRYATWKKAVMKSMGWVTSQSPEGGDPSIFSSLPLGFFIVSSMVMLIGARYISGVPSpecifications
| GK2 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_033214.2 | |
| GK2 | |
| GK2 (NP_149991.2, 1 a.a. ∼ 553 a.a) full-length human protein. | |
| RUO | |
| 2712 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| glycerol kinase 2 | |
| GKP2/GKTA | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |