missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rk1, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 494753-69-4 |
| Linear Formula | C42H70O12 |
| Synonym | (3 beta, 12 beta)-12-Hydroxydammara-20,24-dien-3-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 767.00 |
| Percent Purity | ≥95.0% (HPLC) |
| Grade | Analytical Standard |