missing translation for 'onlineSavingsMsg'
Learn More

Ginsenoside Rg3, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013087
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013087 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013087 Supplier Merck Emd Millipore Supplier No. 6413910MG
Featured Industries: Food and Beverages

Specifications

CAS 14197-60-5
Physical Form Neat
Linear Formula C42H72O13
MDL Number MFCD06410950
Synonym (3 beta,12 beta)-12,20-Dihydroxydammar-24-en-3-yl 2-O-beta-D-glucopyranosyl-beta-D-glucopyranoside; 20-Ginsenoside-Rg3; Rg3
Type Analytical Standard
Quantity 10 mg
Formula Weight 785.01
Percent Purity ≥96% (HPLC)
Grade Analytical Standard