missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ginsenoside Rg1, ≥90% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 22427-39-0 |
| Physical Form | Neat |
| Linear Formula | C42H72O14 |
| MDL Number | MFCD00210293 |
| Synonym | (3 beta,6 alpha,12 beta)-3,12-Dihydroxydammar-24-ene-6,20-diyl bis-beta-D-glucopyranoside; Ginsenoside A2; Ginsenoside g1; Panaxoside A; Panaxoside Rg1; Sanchinoside C1; Sanchinoside Rg1 |
| Type | Analytical Standard |
| Quantity | 5 mg |
| Formula Weight | 801.01 |
| Percent Purity | ≥90% (HPLC) |
| Grade | Analytical Standard |