missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolic acid C15:1, ≥80% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013605
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013605 Supplier Merck Emd Millipore Supplier No. 757415MG

Ginkgolic acid C15:1 is an alkylphenol belonging to the class of ginkgolic acids isolated from Ginkgo biloba leaves. It also shows strong allergenic properties, with carcinogenic and mutagenic activity.

Food and Beverages

Specifications

CAS 22910-60-7
Flash Point 265.5°C - (calculated)
Physical Form Neat
Linear Formula C22H34O3
MDL Number MFCD03093717
Synonym 22:1-omega7-Anacardic acid; 6-[(8 Z)-Pentadecenyl]-salicylic acid; Anacardic acid monoene; Ginkgolic acid I; Romanicardic acid
Type Analytical Standard
Quantity 5 mg
Formula Weight 346.50
Percent Purity ≥80% (HPLC)
Grade Analytical Standard
Show More Show Less