missing translation for 'onlineSavingsMsg'
Learn More

Ginkgolic acid C13:0, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013463
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013463 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013463 Supplier Merck Emd Millipore Supplier No. 4996210MG

Ginkgolic acid C13:0 is a compound obtained from Ginkgo biloba and is effectively used as an antimicrobial agent against S.mutans.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 20261-38-5
Physical Form Neat
Linear Formula C20H32O3
MDL Number MFCD01661602
Synonym 2-Hydroxy-6-tridecylbenzoic acid; 6-Tridecylsalicylic acid; Ginkgoneolic acid
Type Analytical Standard
Quantity 10 mg
Formula Weight 320.47
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less