missing translation for 'onlineSavingsMsg'
Learn More

Gimeracil 98.0+%, TCI America™

Catalog No. C22431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22431G 1 g
C22435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22431G Supplier TCI America Supplier No. C22431G

Chemical Identifiers

CAS 103766-25-2
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.54
MDL Number MFCD08458352
InChI Key ZPLQIPFOCGIIHV-UHFFFAOYSA-N
Synonym 5-Chloro-2,4-dihydroxypyridine, 5-Chloro-2,4-pyridinediol, 5-Chloro-4-hydroxy-2(1H)-pyridinone
PubChem CID 54679224
IUPAC Name 5-chloro-2-hydroxy-1,4-dihydropyridin-4-one
SMILES OC1=CC(=O)C(Cl)=CN1

Specifications

Melting Point 274°C
Color White-Yellow
Quantity 1 g
Formula Weight 145.54
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Gimeracil