accessibility menu, dialog, popup

UserName

OHOHNCl

CAS RN 103766-25-2

IUPAC Name: 5-chloro-2-hydroxy-1,4-dihydropyridin-4-one
Synonyms: 5-chloro-2-hydroxy-1H-pyridin-4-one
Molecular Weight (g/mol): 145.54
Molecular Formula: C5H4ClNO2
InChi Key: ZPLQIPFOCGIIHV-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C(Cl)=CN1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results