Learn More
Gilson™ PIPETMANTM L Multichannel Pipet
GREENER_CHOICE
The PIPETMANTM L Multichannel Pipet is a lightweight multichannel pipet with lockable volume that has been designed to fit naturally in your hand.
Supplier:  Gilson™ FA10011
 View more versions of this product
Catalog No. FA10011G
Get Up to $1,500 in Gilson Products with Purchase
View all promotions
Only null left
Description
Description
  • Gilson–patented volume locking system guarantees no accidental change in volume while pipetting.
  • Adjustable tip ejector provides comfort for right- and left-handed users.
  • Redesigned handle fits naturally in your hand, no matter the size.
  • Unique internal design applies even, equal pressure from either side of the pipette, rather than on the center of the channels like other multichannels do.
  • 2D identification code tracks product reference number, date of manufacture, serial number and maximum volume of pipet.
  • Available in 8–and 12–channel covering a volume range from 0.5 to 300μL.
Specifications
Chemical Identifiers
No
Manual
Ergonomic
Pipetman
Non-sterile
Ergonomic
1 Year
Autoclavable
Reusable
Polypropylene
No
Pipette
1
Certified Quality
Specifications
PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST
Autoclavable
Manual
Pipette Tip 2 to 200 μL
Polypropylene
8
No
Pipette
Ergonomic
1 Year
15.2 cm
32.6 cm
3.9 cm
Certified Quality
No
Yellow
Reusable
Ergonomic
P8x200L
Pipetman
Non-sterile
20 to 200 μL
1
5.98 in.
12.83 in.
1.54 in.
0.81 lb.
Product Suggestions
Product Suggestions