Learn More
Gilson™ PIPETMANTM EasyPackTM Tips
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMANTM EasyPackTM Tips.
$80.82 - $96.38
Specifications
| Filtered | Non-filtered |
|---|---|
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Format | Bagged |
| Tip Style | Universal |
| Catalog Number | Mfr. No. | Volume (Metric) | Format | Quantity | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Volume (Metric) | Format | Quantity | Price | Quantity | |||
|
F161210G
|
Gilson™
F161210 |
1 to 10 mL | Bagged | 200 (4x50) |
Pack of 200 Each for $87.04
|
|
|||
|
F161111G
|
Gilson™
F161111 |
100 to 1200 μL | Bagged | 200 |
Pack of 1000 Each for $87.04
|
|
|||
|
F1614501G
|
Gilson™
F1614501 |
0.1 to 20 μL | Bagged | 1000 |
Pack of 1000 Each for $87.04
|
|
|||
|
F161451G
|
Gilson™
F161451 |
0.1 to 20 μL | Bagged | 200 |
Pack of 1000 Each for $96.38
|
|
|||
|
F1616301G
|
Gilson™
F1616301 |
0.1 to 10 μL | Bagged | 1000 |
Pack of 1000 Each for $80.82
|
|
|||
|
F161571G
|
Gilson™
F161571 |
1 to 5 mL | Bagged | 334 |
Pack of 1002 Each for $93.26
|
|
|||
|
F161631G
|
Gilson™
F161631 |
0.1 to 10 μL | Bagged | 1000 (5x200) |
Pack of 1000 Each for $94.82
|
|
|||
|
F1616701G
|
Gilson™
F1616701 |
100 to 1000 μL | Bagged | 1000 |
Pack of 1000 Each for $80.82
|
|
|||
|
F161671G
|
Gilson™
F161671 |
100 to 1000 μL | Bagged | 1000 (5x200) |
Pack of 1000 Each for $94.82
|
|
|||
|
F1617301G
|
Gilson™
F1617301 |
20 to 300 μL | Bagged | 1000 |
Pack of 1000 Each for $80.82
|
|
|||
|
F161931G
|
Gilson™
F161931 |
2 to 200 μL | Bagged | 1000 (5x200) |
Pack of 1000 Each for $94.82
|
|
|||
|
F1619301G
|
Gilson™
F1619301 |
2 to 200 μL | Bagged | 1000 |
Pack of 1000 Each for $80.82
|
|
|||
|
F161731G
|
Gilson™
F161731 |
20 to 300 μL | Bagged | 1000 (5x200) |
Pack of 1000 Each for $94.82
|
|
|||
Description
- Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
- Manufactured in a cleanroom and hence no risk of contamination.
- Full traceability from the mold to the bench guaranteed.
- Volume graduations make it easy to visually check the volume aspirated.
- Requires low tip-mounting force.
- Easy identification with color-coded packaging.
- Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
- Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
Specifications
| Non-filtered | |
| Autoclavable | |
| Universal | |
| Gilson Pipetman | |
| Polypropylene |
| Non-sterile | |
| Bagged | |
| Pipet Tips | |
| Yes |