Learn More
Gilson™ MICROMANTM E Positive Displacement Pipette
GREENER_CHOICE
The ergonomically designed MICROMANTM E Positive Displacement Pipette, along with disposable capillary piston (CP) tips, provide the highest precision when pipetting viscous, non-aqueous liquids such as oils, blood, glycerol, and more, while protecting the pipette from contamination.
Supplier:  Gilson™ FD10003
 View more versions of this product
Catalog No. FD10003G
Get Up to $1,500 in Gilson Products with Purchase
View all promotions
Only null left
Description
Description
MICROMANTM E positive displacement pipettes with capillary pistons offer highest precision when pipetting problem liquids such as oils, cosmetics, blood, glycerol, radioactive samples, RNA, DNA, and more.
  • Ergonomic design to increase pipetting performance.
  • Patented QuickSnap feature for Capillary Piston fitting System.
  • Volume securing button to reduce risk of accidental volume change during pipetting cycles.
  • Comfortable and easy to use with new handle shape and added finger hook.
  • Fully visible volumeter.
  • Large pushbutton for allows easy aspirate and dispense.
  • For complete and rapid pipetting of viscous, volatile, dense, hot or cold liquids.
  • Avoids leaks when pipetting volatile (high vapor pressure) liquids.
  • Avoid cross-contamination or residual sample carry-over.
  • For tips, see F148412G or F148413G.
Specifications
Chemical Identifiers
Autoclavable
1
For pipetting viscous, volatile, and problem liquids
Certified Quality
Polypropylene
Positive Displacement Pipette
1
Specifications
CP Tips
Blue
M50E
Positive Displacement Pipette
For pipetting viscous, volatile, and problem liquids
4.25 in.
1.42 in.
Autoclavable
Polypropylene
1
20 to 50 μL
1
12.68 in.
Certified Quality
Product Suggestions
Product Suggestions